Learn More
Medchemexpress LLC Guanylate-binding protein 2 | >=96.0% | 100 UG
SDP
Catalog No. 5000294360
Encompass_Preferred
Recombinant human guanylate-binding protein 2 expressed in HEK293 cells with a C-terminal His tag, supplied as a lyophilized powder. The protein corresponds to residues M1-C588, has an apparent molecular weight of ~60-75 kDa due to glycosylation, and is characterized to ≥96.0% purity by reducing SDS-PAGE. Formulated in PBS, pH 7.4 with 8% trehalose; endotoxin <1 EU/μg; reconstitute to ≥100 μg/mL and store frozen for long term.
  • Lyophilized recombinant human protein, ready for reconstitution.
  • High purity (≥96.0%) suitable for biochemical assays.
  • Low endotoxin level (<1 EU/μg) for cellular applications.
  • Formulated with trehalose for stability during storage.
  • Recommended reconstitution concentration of at least 100 μg/mL.
  • Stable long-term when aliquoted and frozen at -20°C or -80°C.
Catalog No. 50-002-94360 Supplier Medchemexpress LLC Supplier No. HYP75787100UG
Recombinant human guanylate-binding protein 2 expressed in HEK293 cells with a C-terminal His tag, supplied as a lyophilized powder. The protein corresponds to residues M1-C588, has an apparent molecular weight of ~60-75 kDa due to glycosylation, and is characterized to ≥96.0% purity by reducing SDS-PAGE. Formulated in PBS, pH 7.4 with 8% trehalose; endotoxin <1 EU/μg; reconstitute to ≥100 μg/mL and store frozen for long term.
  • Lyophilized recombinant human protein, ready for reconstitution.
  • High purity (≥96.0%) suitable for biochemical assays.
  • Low endotoxin level (<1 EU/μg) for cellular applications.
  • Formulated with trehalose for stability during storage.
  • Recommended reconstitution concentration of at least 100 μg/mL.
  • Stable long-term when aliquoted and frozen at -20°C or -80°C.