Learn More
Bachem Guanylin (human)
SDP
Supplier:  Bachem 4030693.1000
Encompass_Preferred
Guanylin (human), H-Pro-Gly-Thr-Cys-Glu-Ile-Cys-Ala-Tyr-Ala-Ala-Cys-Thr-Gly-Cys-OH, 183200-12-6, C58H87N15O21S4, Mr 1458.68, 1mg. Endogenous intestinal guanylate cyclase activator.
Catalog No. 50-259-683