Learn More
CPC Scientific H-Pro-Gly-Thr-Cys-Glu-Ile-Cys-Ala-Tyr-Ala-Ala-Cys-Thr-Gly-Cys-OH (trifluoroacetate salt)
(Cys4 and 12 bridge, Cys7 and 15 bridge) 0.5MG
SDP
Catalog No. 502109410
Encompass
Intestinal guanylate cyclase activator that is isolated from rat intestine and activates intestinal guanylate cyclase in a human colon carcinoma-derived cell line (T84). It resembles the heat-stable endotoxins produced by E. coli.SEQUENCE: H-Pro-Gly-Thr-Cys-Glu-Ile-Cys-Ala-Tyr-Ala-Ala-Cys-Thr-Gly-Cys-OH (trifluoroacetate salt)(Cys4 and 12 bridge, Cys7 and 15 bridge)ONE-LETTER SEQUENCE: PGTCEICAYAACTGC (C4&C12 bridge, C7&C15 bridge)MOLECULAR FORMULA: C58H87N15O21S4MOLECULAR WEIGHT: 1458.68STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [183200-12-6]SYNONYMS:RESEARCH AREA: GastrointestinalREFERENCES:R.C. Wiegand et al., FEBS Letters, 311, 150 (1992)F.J.de Sauvage et al., Proc. Natl. Acad. Sci. USA, 89, 9089 (1992)SKU(s): GUAN-001A, GUAN-001B
Catalog No. 50-210-9410 Supplier CPC Scientific Supplier No. GUAN001A
Add to Cart
Add to Cart
Intestinal guanylate cyclase activator that is isolated from rat intestine and activates intestinal guanylate cyclase in a human colon carcinoma-derived cell line (T84). It resembles the heat-stable endotoxins produced by E. coli.SEQUENCE: H-Pro-Gly-Thr-Cys-Glu-Ile-Cys-Ala-Tyr-Ala-Ala-Cys-Thr-Gly-Cys-OH (trifluoroacetate salt)(Cys4 and 12 bridge, Cys7 and 15 bridge)ONE-LETTER SEQUENCE: PGTCEICAYAACTGC (C4&C12 bridge, C7&C15 bridge)MOLECULAR FORMULA: C58H87N15O21S4MOLECULAR WEIGHT: 1458.68STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [183200-12-6]SYNONYMS:RESEARCH AREA: GastrointestinalREFERENCES:R.C. Wiegand et al., FEBS Letters, 311, 150 (1992)F.J.de Sauvage et al., Proc. Natl. Acad. Sci. USA, 89, 9089 (1992)SKU(s): GUAN-001A, GUAN-001B