Learn More
Bachem Guanylin (mouse, rat)
SDP
Supplier:  Bachem 4030694.0500
Encompass_Preferred
Guanylin (mouse, rat) , H-Pro-Asn-Thr-Cys-Glu-Ile-Cys-Ala-Tyr-Ala-Ala-Cys-Thr-Gly-Cys-OH, 144940-98-7, C60H90N16O22S4, Mr 1515.73, 0.5mg. Guanylin activates the guanylate cyclase in the intestine to stimulate the production of cGMP, which in turn causes a secretory diarrhoea. Therefore, this peptide may play a role in regulating fluid and electrolyte absorption in intestines.
Catalog No. 50-382-414