Learn More
Enzo Life Sciences Guanylin (rat, mouse) (0.1mg). CAS: 144940-98-7
SDP
Supplier:  Enzo Life Sciences ALX157011MC01
Encompass_Preferred
Endogenous intestinal guanylyl cyclase activator. Formula: C60H90N16O22S4. MW: 1515.7. Purity: ≥99%. Appearance: White to off-white powder. Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-7821