Learn More
ABclonal Technology Guanylyl Cyclase beta 1 (GUCY1B3) Rabbit pAb
SDP
Supplier:  ABclonal Technology A152761000UL
Encompass_Preferred
This gene encodes the beta subunit of the soluble guanylate cyclase (sGC), which catalyzes the conversion of GTP (guanosine triphosphate) to cGMP (cyclic guanosine monophosphate). The encoded protein contains an HNOX domain, which serves as a receptor for ligands such as nitric oxide, oxygen and nitrovasodilator drugs. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-06376