Learn More
Medchemexpress LLC Guanylyl imidodiphosphate lithium | 64564-03-0 | 98.0% | C10H13Li4N6O13P3 | 10MG
SDP
Catalog No. 5000259102
Encompass_Preferred
Guanylyl imidodiphosphate lithium (Gpp(NH)p) is a non-hydrolyzable guanosine triphosphate analogue used to probe G-protein-mediated signaling. It mimics GTP binding while resisting enzymatic hydrolysis, enabling sustained activation of G proteins and increased adenylate cyclase activity in biochemical and pharmacological studies.
  • Non-hydrolyzable GTP analogue for stable G-protein activation.
  • Used to increase adenylate cyclase activity in signaling assays.
  • Provided as a solid tetralithium salt for biochemical use.
  • High purity (98.0%) suitable for research applications.
  • Stable under frozen storage to preserve activity.
Catalog No. 50-002-59102 Supplier Medchemexpress LLC Supplier No. HY13716710MG
Add to Cart
Add to Cart
Guanylyl imidodiphosphate lithium (Gpp(NH)p) is a non-hydrolyzable guanosine triphosphate analogue used to probe G-protein-mediated signaling. It mimics GTP binding while resisting enzymatic hydrolysis, enabling sustained activation of G proteins and increased adenylate cyclase activity in biochemical and pharmacological studies.
  • Non-hydrolyzable GTP analogue for stable G-protein activation.
  • Used to increase adenylate cyclase activity in signaling assays.
  • Provided as a solid tetralithium salt for biochemical use.
  • High purity (98.0%) suitable for research applications.
  • Stable under frozen storage to preserve activity.