Learn More
Medchemexpress LLC Guar gum | 9000-30-0 | MFCD00131250 | 98.0% | 50,000-8,000,000 Da | (C6H10O5)n·H2O | 250 G
SDP
Catalog No. 5000283085
Encompass_Preferred
Guar gum is a natural galactomannan polysaccharide used as a pharmaceutical excipient and functional polymer. It delivers thickening, emulsifying, binding, and gelling functionality, dissolves readily in cold water, shows wide pH stability, and is biodegradable - making it suitable for use in drug formulations and other industrial applications.
  • Provides thickening, emulsifying, binding, and gelling properties.
  • Quickly soluble in cold water.
  • Wide pH stability across formulation conditions.
  • Biodegradable and film-forming.
  • Suitable as an excipient for thickening and suspending in formulations.
  • Offered in multiple pack sizes for laboratory and production use.
Catalog No. 50-002-83085 Supplier Medchemexpress LLC Supplier No. HYW145516250G
Guar gum is a natural galactomannan polysaccharide used as a pharmaceutical excipient and functional polymer. It delivers thickening, emulsifying, binding, and gelling functionality, dissolves readily in cold water, shows wide pH stability, and is biodegradable - making it suitable for use in drug formulations and other industrial applications.
  • Provides thickening, emulsifying, binding, and gelling properties.
  • Quickly soluble in cold water.
  • Wide pH stability across formulation conditions.
  • Biodegradable and film-forming.
  • Suitable as an excipient for thickening and suspending in formulations.
  • Offered in multiple pack sizes for laboratory and production use.