Learn More
Medchemexpress LLC Guar gum | 9000-30-0 | MFCD00131250 | 98.0% | 50,000-8,000,000 g/mol | 50 G
SDP
Catalog No. 5000283548
Encompass_Preferred
Guar gum is a natural galactomannan polysaccharide extracted from the endosperm of Cyamopsis tetragonolobus seeds. Supplied as a white to off-white powder, it is used as a pharmaceutical excipient and functional polymer in drug formulation to provide thickening, emulsifying, binding, gelling, and suspending properties and to improve stability, solubility, and processability.
  • High purity (98.0%).
  • White to off-white powder with quick solubility in cold water.
  • Functions as thickener, emulsifier, binder, and gelling agent.
  • Suitable for drug delivery and formulation development.
  • Available in multiple lab-scale package sizes.
Catalog No. 50-002-83548 Supplier Medchemexpress LLC Supplier No. HYW14551650G
Guar gum is a natural galactomannan polysaccharide extracted from the endosperm of Cyamopsis tetragonolobus seeds. Supplied as a white to off-white powder, it is used as a pharmaceutical excipient and functional polymer in drug formulation to provide thickening, emulsifying, binding, gelling, and suspending properties and to improve stability, solubility, and processability.
  • High purity (98.0%).
  • White to off-white powder with quick solubility in cold water.
  • Functions as thickener, emulsifier, binder, and gelling agent.
  • Suitable for drug delivery and formulation development.
  • Available in multiple lab-scale package sizes.