Learn More
Medchemexpress LLC Guar gum | 9000-30-0 | MFCD00131250 | 98.0% | 50,000-8,000,000 g/mol | (C6H10O5)n | 1 KG
SDP
Catalog No. 5000283087
Encompass_Preferred
Guar gum is a high-molecular-weight galactomannan polysaccharide used as a multifunctional pharmaceutical excipient and formulation additive. Supplied as a powdered material (CAS 9000-30-0) with a stated purity of 98.0%, it is employed to modify viscosity, stabilize dispersions, and improve tablet cohesion in drug-delivery and related applications.
  • Acts as a thickener and viscosity modifier in aqueous formulations.
  • Functions as an emulsifier and stabilizer for suspensions and emulsions.
  • Provides binding and film-forming properties for solid dosage forms.
  • Hydrates rapidly and dissolves in cold water for easy processing.
  • Biodegradable and broadly stable across common pH ranges.
Catalog No. 50-002-83087 Supplier Medchemexpress LLC Supplier No. HYW1455161KG
Guar gum is a high-molecular-weight galactomannan polysaccharide used as a multifunctional pharmaceutical excipient and formulation additive. Supplied as a powdered material (CAS 9000-30-0) with a stated purity of 98.0%, it is employed to modify viscosity, stabilize dispersions, and improve tablet cohesion in drug-delivery and related applications.
  • Acts as a thickener and viscosity modifier in aqueous formulations.
  • Functions as an emulsifier and stabilizer for suspensions and emulsions.
  • Provides binding and film-forming properties for solid dosage forms.
  • Hydrates rapidly and dissolves in cold water for easy processing.
  • Biodegradable and broadly stable across common pH ranges.