Learn More
Medchemexpress LLC Guar gum | 9000-30-0 | MFCD00131250 | 98.0% | 50,000-8,000,000 g/mol | Polymeric - repeating unit (C6H10O5)n (galactomannan; no single molecular formula) | 25 G
SDP
Catalog No. 5000283754
Encompass_Preferred
Guar gum is a high-molecular-weight galactomannan polysaccharide supplied as a white to off-white powder. It is used as a pharmaceutical excipient and formulation aid for its thickening, suspending, emulsifying, binding, and gelling properties; typical purity is 98% and it is available in multiple pack sizes with defined storage recommendations.
  • Provides thickening and viscosity enhancement in aqueous formulations.
  • Acts as a suspending agent to improve dispersion stability.
  • Offers emulsifying and binding properties for solid and liquid dosage forms.
  • Water-dispersible powder for straightforward formulation handling.
  • White to off-white appearance suitable for many applications.
  • Long shelf life when stored per recommended conditions.
Catalog No. 50-002-83754 Supplier Medchemexpress LLC Supplier No. HYW14551625G
Guar gum is a high-molecular-weight galactomannan polysaccharide supplied as a white to off-white powder. It is used as a pharmaceutical excipient and formulation aid for its thickening, suspending, emulsifying, binding, and gelling properties; typical purity is 98% and it is available in multiple pack sizes with defined storage recommendations.
  • Provides thickening and viscosity enhancement in aqueous formulations.
  • Acts as a suspending agent to improve dispersion stability.
  • Offers emulsifying and binding properties for solid and liquid dosage forms.
  • Water-dispersible powder for straightforward formulation handling.
  • White to off-white appearance suitable for many applications.
  • Long shelf life when stored per recommended conditions.