Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guar gum | 9000-30-0 | MFCD00131250 | 98.0% | Not determined - polymer; no single molecular weight | Not applicable - polysaccharide polymer (no single chemical formula) | 100 G
SDP

Catalog No. 5000283086
Encompass_Preferred

Guar gum is a plant-derived galactomannan polysaccharide provided as a white to off-white powder for use as a pharmaceutical excipient in formulation and drug delivery research. It offers thickening, binding, gelling, emulsifying, and suspending functionality with good film-forming ability and biodegradability.

  • Appearance: white to off-white powder.
  • Purity: 98.0%.
  • CAS number: 9000-30-0.
  • Function: thickener, binder, gelling, emulsifying, and suspending agent.
  • Solubility: rapid solubility in cold water; wide pH stability.
  • Physical property: film-forming and biodegradable.
  • Storage guidance: powder storage at -20°C or 4°C with extended shelf life when frozen.

Catalog No. 50-002-83086 Supplier Medchemexpress LLC Supplier No. HYW145516100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guar gum is a plant-derived galactomannan polysaccharide provided as a white to off-white powder for use as a pharmaceutical excipient in formulation and drug delivery research. It offers thickening, binding, gelling, emulsifying, and suspending functionality with good film-forming ability and biodegradability.

  • Appearance: white to off-white powder.
  • Purity: 98.0%.
  • CAS number: 9000-30-0.
  • Function: thickener, binder, gelling, emulsifying, and suspending agent.
  • Solubility: rapid solubility in cold water; wide pH stability.
  • Physical property: film-forming and biodegradable.
  • Storage guidance: powder storage at -20°C or 4°C with extended shelf life when frozen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.