Learn More
Bioworld GUAR GUM SEPAROPOREAGAROS
Catalog No. 50197799
Encompass
GUAR GUM SEPAROPORE (AGAROSE) 4B-CL: Guar gum is a Galactomannan polymer derived from the seeds of the legume plant Cyamopsis tetragonolobus.Guar gum based affinity chromatography is used for the purification of galactose binding prot...
Catalog No. 50-197-799 Supplier Bioworld Supplier No. 201810523
GUAR GUM SEPAROPORE (AGAROSE) 4B-CL: Guar gum is a Galactomannan polymer derived from the seeds of the legume plant Cyamopsis tetragonolobus.Guar gum based affinity chromatography is used for the purification of galactose binding prot...