Learn More
Bioworld GUAR GUM SEPAROPOREAGAROS
Supplier:  Bioworld 201810521
Encompass
GUAR GUM SEPAROPORE (AGAROSE) 4B-CL: Guar gum is a Galactomannan polymer derived from the seeds of the legume plant Cyamopsis tetragonolobus.Guar gum based affinity chromatography is used for the purification of galactose binding prot...
Catalog No. 50-197-797