Learn More
Tosoh Bioscience LLC GUARD 4.6MM SUPERHZ1000-4000
Supplier:  Tosoh Bioscience LLC 0019314
Encompass
Column, Guard; Tosoh; TSKgel SuperHZ 4000; Media: Polymer; Stainless-steel; Particle: 4u m; For gel permeation chromatography; 4.6mm I.D. x 2cm length
Catalog No. 50-851-547