Learn More
Thermo Scientific GUARD COLUMN C18 MG 2/PKG. 3
Supplier:  Thermo Scientific 8812305
Encompass
Drop-in guard cartridges, MG, C18, 4x10mm, 2/pk - fits guard column holder Cat No. NC0527629 (No. 88-12415)
Catalog No. NC0248127