Learn More
Microsolv Technology Corporation Guard Column Cartridges, RP C18 ABX, 3um, 100A. 2.1mm ID x 10mm Length. 5/PK.
Catalog No. 502270254
Encompass_Preferred
These Guard Column Cartridges are packed with 3µm, Cogent RP C18 ABX™ Stationary Phase. They are specially end capped ( bulky ), with traditional bonding of an 18 carbon ligand to the highest purity silica available.Designed to protect your 2.1mm ID, RP C18 ABX, Analytical Columns from particulate and contaminants that will adsorb to it. They are packed in Stainless Steel Tubes and have Stainless Steel Frits that are pressed into PEEK Frit Caps. These Guard Column Cartridges must be used with a Guard Column Holder.
This item is not returnable. View return policy
Catalog No. 50-227-0254 Supplier Microsolv Technology Corporation Supplier No. 85318PG1
These Guard Column Cartridges are packed with 3µm, Cogent RP C18 ABX™ Stationary Phase. They are specially end capped ( bulky ), with traditional bonding of an 18 carbon ligand to the highest purity silica available.Designed to protect your 2.1mm ID, RP C18 ABX, Analytical Columns from particulate and contaminants that will adsorb to it. They are packed in Stainless Steel Tubes and have Stainless Steel Frits that are pressed into PEEK Frit Caps. These Guard Column Cartridges must be used with a Guard Column Holder.