Learn More
Microsolv Technology Corporation Guard Column Cartridge, PEEK, 3.0mm ID, 8mm active length. Frits sold separately. 1 EA.
Catalog No. 502271555
Encompass_Preferred
This Guard Column Cartridge (tube) is 3.0mm ID. Extruded from Victrex PEEK, no chemical treatment is done and the interior mirror like finish is provided through our very careful extrusion techniques. Frits are sold separately.
This item is not returnable. View return policy
Catalog No. 50-227-1555 Supplier Microsolv Technology Corporation Supplier No. 49330GC2
This Guard Column Cartridge (tube) is 3.0mm ID. Extruded from Victrex PEEK, no chemical treatment is done and the interior mirror like finish is provided through our very careful extrusion techniques. Frits are sold separately.