Learn More
Microsolv Technology Corporation Guard Column Cartridges, RP C18 PE,5um, 100A. 2.1mm ID x 10mm Length. 5/PK.
Catalog No. 502270236
Encompass_Preferred
These Guard Column Cartridges are packed with 5µm, Cogent RP C18 PE™ Stationary Phase. Double end capped (fully), it is bonded with an 18 carbon aliphatic hydrocarbon, with an Amide group embedded in the middle of the ligand, using the highest purity silica available.Designed to protect your 2.1mm ID, RP C18 PE, Analytical Columns from particulate and contaminants that will adsorb to it. They are packed in Stainless Steel Tubes and have Stainless Steel Frits that are pressed into PEEK Frit Caps. These Guard Column Cartridges must be used with a Guard Column Holder.
This item is not returnable. View return policy
Catalog No. 50-227-0236 Supplier Microsolv Technology Corporation Supplier No. 84518PG1
These Guard Column Cartridges are packed with 5µm, Cogent RP C18 PE™ Stationary Phase. Double end capped (fully), it is bonded with an 18 carbon aliphatic hydrocarbon, with an Amide group embedded in the middle of the ligand, using the highest purity silica available.Designed to protect your 2.1mm ID, RP C18 PE, Analytical Columns from particulate and contaminants that will adsorb to it. They are packed in Stainless Steel Tubes and have Stainless Steel Frits that are pressed into PEEK Frit Caps. These Guard Column Cartridges must be used with a Guard Column Holder.