Learn More
Microsolv Technology Corporation Guard Column Cartridges, RP C18, 10um, 100A. 4.0mm ID x 10mm Length. 5/PK.
Catalog No. 502270195
Encompass_Preferred
These Guard Column Cartridges are packed with 10µm, Cogent RP C18™ Stationary Phase. They are double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available. Designed to protect your 3.0, 4.0 or 4.6mm ID, RP C18, Analytical Columns from particulate and contaminants that will adsorb to it. They are packed in Stainless Steel Tubes and have Stainless Steel Frits that are pressed into PEEK Frit Caps. These Guard Column Cartridges must be used with a Guard Column Holder.
This item is not returnable. View return policy
Catalog No. 50-227-0195 Supplier Microsolv Technology Corporation Supplier No. 68018PG2
These Guard Column Cartridges are packed with 10µm, Cogent RP C18™ Stationary Phase. They are double end capped ( fully ), with traditional bonding of an 18 carbon silane to the highest purity silica available. Designed to protect your 3.0, 4.0 or 4.6mm ID, RP C18, Analytical Columns from particulate and contaminants that will adsorb to it. They are packed in Stainless Steel Tubes and have Stainless Steel Frits that are pressed into PEEK Frit Caps. These Guard Column Cartridges must be used with a Guard Column Holder.