Learn More
Microsolv Technology Corporation Guard Column Holder, 10mm long Cogent RP, NP or IEX Guard. Accepts 1.0-4.0mm ID Cartridges. 1 EA.
Catalog No. 502270211
Encompass_Preferred
Guard Column Holder, 10mm long Cogent RP, NP or IEX Guard. Accepts 1.0-4.0mm ID Cartridges. 1 EA.
This item is not returnable. View return policy
Catalog No. 50-227-0211 Supplier Microsolv Technology Corporation Supplier No. 83000GH
Guard Column Holder, 10mm long Cogent RP, NP or IEX Guard. Accepts 1.0-4.0mm ID Cartridges. 1 EA.