Learn More
Microsolv Technology Corporation Guard Column Holder, Semi Prep, Cogent RP, NP or IEX Guard. Accepts 10mm x 10mm ID Prep Cartridges. 1 EA.
Supplier:  Microsolv Technology Corporation 83000SPGH
Encompass_Preferred
This Guard Column Holder accepts Cogent RP, Cogent NP or Cogent IEX Prep Guard Columns (Cartridges) that are 10mm in Internal Diameter and are 10mm Long which are sold separately. Stainless Steel Body and Frits, this system is a great way to keep your 10mm Prep Columns (all lengths) from being contaminated by your samples or impurities that might adsorb onto the Column and keep out some of the undissolved particles.
Catalog No. 50-227-0214