Learn More
DSI LLC GUARD COLUMN INSERT STAINLESS
Catalog No. 50123097
Encompass_Preferred
ZirChrom-WCX. Weak Cation-exchange. 300 Angstrom. Useful for protein chromatography in the cation-exchange mode. Stable over a pH range of 1-10.
Catalog No. 50-123-097 Supplier DSI LLC Supplier No. ZR04G40
ZirChrom-WCX. Weak Cation-exchange. 300 Angstrom. Useful for protein chromatography in the cation-exchange mode. Stable over a pH range of 1-10.