Learn More
DSI LLC GUARD COLUMN INSERT STAINLESS
Catalog No. 50123050
Encompass_Preferred
DiamondBond-C18 eliminates the shortcomings of conventional ODS-silica columns by combining patented surface technology with ZirChrom's state-of-the-art particles. Carbon-clad zirconia, stable at all levels of pH.
Catalog No. 50-123-050 Supplier DSI LLC Supplier No. DB01G40
DiamondBond-C18 eliminates the shortcomings of conventional ODS-silica columns by combining patented surface technology with ZirChrom's state-of-the-art particles. Carbon-clad zirconia, stable at all levels of pH.