Learn More
Microsolv Technology Corporation Guard Column Kit, PEEK, 3.0mm ID, 8mm active length. 1 EA.
Catalog No. 502271554
Encompass_Preferred
This Guard Column Hardware Kit includes one 3.0mm ID Column Tube, two frits and the Guard Column Holder. All components, including the Frits are extruded from Victrex PEEK. No chemical treatment is done to the tubes and the interior mirror like finish is provided through our very careful extrusion techniques. Carbon is added to black components.
This item is not returnable. View return policy
Catalog No. 50-227-1554 Supplier Microsolv Technology Corporation Supplier No. 49330GC1
This Guard Column Hardware Kit includes one 3.0mm ID Column Tube, two frits and the Guard Column Holder. All components, including the Frits are extruded from Victrex PEEK. No chemical treatment is done to the tubes and the interior mirror like finish is provided through our very careful extrusion techniques. Carbon is added to black components.