Learn More
Mac-Mod Avantor Prevail Amino, 110 Å, 5 µm, 4.6 x 7.5 mm Guard Cartridges (3/pk)
SDP
Catalog No. 5000153109
Encompass
Avantor Prevail HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. Avantor Prevail HPLC columns are silica based except for Prevail Carbohydrate ES. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes. Avantor Prevail HPLC columns are stable from highly organic to highly aqueous. Avantor Prevail HPLC columns are specialty phases for specific applications. Avantor Prevail HPLC columns provide excellent sensitivity with microbore and ELSD applications.
Catalog No. 50-001-53109 Supplier Mac-Mod Supplier No. 99290N
Avantor Prevail HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. Avantor Prevail HPLC columns are silica based except for Prevail Carbohydrate ES. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes. Avantor Prevail HPLC columns are stable from highly organic to highly aqueous. Avantor Prevail HPLC columns are specialty phases for specific applications. Avantor Prevail HPLC columns provide excellent sensitivity with microbore and ELSD applications.