Learn More
Mac-Mod HydroBond AQ C8, 100 Å, 3 µm, 3.2 x 10 mm Guard Cartridges (5/pk)
SDP
Catalog No. 5000150599
Encompass
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-001-50599 Supplier Mac-Mod Supplier No. MMHB832203
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.