Learn More
Mac-Mod HydroBond AQ C8, 100 Å, 5 µm, 3.2 x 10 mm Guard Cartridges (5/pk)
SDP
Catalog No. 5000151837
Encompass
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.
Catalog No. 50-001-51837 Supplier Mac-Mod Supplier No. MMHB832205
Hydrobond is designed for polar retention of highly polar analytes in RPLC. These columns provide reproducible results, column to column, lot to lot.