Learn More
Mac-Mod Avantor Apex ODS, 100 Å, 5 µm, 4.6 x 7.5 mm Guard Cartridges (3/pk)
SDP
Catalog No. 5000143725
Encompass
Avantor Apex and Avantor Apex ll phases are recommended for routine analyses. Conventional 100 Å pore size spherical silica particle column. Narrow particle size distribution.
Catalog No. 50-001-43725 Supplier Mac-Mod Supplier No. FH13102/N / 5169664/N
Avantor Apex and Avantor Apex ll phases are recommended for routine analyses. Conventional 100 Å pore size spherical silica particle column. Narrow particle size distribution.