Learn More
Mac-Mod Avantor Apollo C18, 100 Å, 5 µm, 3.0; 4.6 x 7.5 mm Guard Cartridges (3/pk)
SDP
Catalog No. 5000144280
Encompass
Avantor Apollo HPLC columns use high purity, base deactivated silica for powerful separations
Catalog No. 50-001-44280 Supplier Mac-Mod Supplier No. 96454/N / 5130575/N
Avantor Apollo HPLC columns use high purity, base deactivated silica for powerful separations