Learn More
Checkers Guard Dog™ ADA Cable Protector
Catalog No. 17456133 Shop All Justrite Safety Group Products
Change view
Click to view available options
Length (Metric):
1.01 m
Catalog No. Length (Metric)
17-456-133 1.01 m
1 options
Catalog No. 17-456-133 Supplier Checkers Supplier No. GD1X75O/B
Add to Cart
Add to Cart
1-Channel Low-Profile ADA Cable Protector for .75 in. Lines.
  • Protects cables and hoses up to .75 in. outside dia.
  • Dog-Bone™ Connectors to extend to any desired length
  • Reinforced hinged lid for easy cable placement
  • With built-in ADA/DDA Accessibility Ramps
  • Patented 5-bar tread surface for maximum traction
  • Modular interlocking design for easy setup
Specifications
Type Cable Tie
Material Plastic
Color Orange
Length (Metric) 1.01 m