Description
- 5-Channel General Purpose ADA/DDA Guard Dog Cable Protector has built-in ADA accessibility ramps to provide a safer crossing for vehicle and pedestrian traffic while protecting valuable electrical cables and hose lines
- Because the ramps are built-in, there is no need for separate connectors
- Another unique feature of this product is that it interconnects directly to a 5-channel standard ramp Guard Dog cable protector
- This modular interlocking design is available in lightweight 18 in sections and it includes a recessed carrying handle on the underside of the protector
- Ideal for use in amusement parks, entertainment venues, sporting events, and most public, commercial, industrial, construction, and utility applications
Specifications
Specifications
| Color | Black |
| Product Type | Cable Protector |
| Dimensions (L x W x H) | 39.5 x 19.5 x 2.13 in. |
| Material | Polyurethane |