Learn More
YMC America, Inc GUARD ODS-A 5UM 120A 10MM ID
SDP
Catalog No. 501107765
Encompass
Cartridge Columns, ODS-A, 5um, 120A, 10x20mm ID, 2/pk. *Requires holder part number XPCHSPW2*
Catalog No. 50-110-7765 Supplier YMC America, Inc Supplier No. AA12S050120CCN
Cartridge Columns, ODS-A, 5um, 120A, 10x20mm ID, 2/pk. *Requires holder part number XPCHSPW2*