Learn More
Lakeside Manufacturing Inc Guard Rail Utility Cart, 2 Shelf: 18" x 27", Heavy-Duty 700-Lb. Capacity
Catalog No. 502884513
Encompass_Preferred
Guard Rail Utility Carts by Lakeside Manufacturing eliminate spills and breakage during transport of medical supplies that could easily tumble off the shelves. The Lakeside 725 Guard Rail Utility Cart with (2) shelves measuring 18" x 27" has a weight capacity of 700-Lb. This utility cart's front rails on all three of its shelves prevent items from falling during transport. This Lakeside cart's all-swivel casters provide ultimate maneuverability, while its tubular push handle comfortably guides the cart with ease. This cart's all-welded construction for a stronger structure and easier-to-clean surface. This Lakeside cart's stainless steel is easy to clean and sanitize, while its bumpers on legs and handles protect walls and furniture. Customize your design with Lakeside cart experts. Superior lead times. Made in the USA.
Catalog No. 50-288-4513 Supplier Lakeside Manufacturing Inc Supplier No. 725
Guard Rail Utility Carts by Lakeside Manufacturing eliminate spills and breakage during transport of medical supplies that could easily tumble off the shelves. The Lakeside 725 Guard Rail Utility Cart with (2) shelves measuring 18" x 27" has a weight capacity of 700-Lb. This utility cart's front rails on all three of its shelves prevent items from falling during transport. This Lakeside cart's all-swivel casters provide ultimate maneuverability, while its tubular push handle comfortably guides the cart with ease. This cart's all-welded construction for a stronger structure and easier-to-clean surface. This Lakeside cart's stainless steel is easy to clean and sanitize, while its bumpers on legs and handles protect walls and furniture. Customize your design with Lakeside cart experts. Superior lead times. Made in the USA.