Learn More
Grainger Porta-King Guard Tower, Welded Steel, Gray, 1 ea
Catalog No. 502775301
Encompass_Preferred
This guard tower provides a durable and secure solution for personnel monitoring and site access control. Constructed from heavy-duty welded steel with an acrylic enamel gray finish, this unit is designed for long-term use in various environments. The structure features high-quality insulation in both walls and ceilings, and the floor is finished with maintenance-free aluminum diamond plate for added durability. The system arrives unassembled and includes factory-installed electrical components ready for on-site connection.
  • Features corrosion-resistant welded steel construction
  • Includes durable aluminum diamond plate floor finish
  • Equipped with high-security steel or anodized aluminum doors
  • Offers R-10 wall insulation and R-19 ceiling insulation
  • Includes factory-installed lighting and electrical systems
  • Features a solar-reflective, waterproof roof membrane
Catalog No. 50-277-5301 Supplier Grainger Supplier No. 18C843
Add to Cart
Add to Cart
Request a Quote
This guard tower provides a durable and secure solution for personnel monitoring and site access control. Constructed from heavy-duty welded steel with an acrylic enamel gray finish, this unit is designed for long-term use in various environments. The structure features high-quality insulation in both walls and ceilings, and the floor is finished with maintenance-free aluminum diamond plate for added durability. The system arrives unassembled and includes factory-installed electrical components ready for on-site connection.
  • Features corrosion-resistant welded steel construction
  • Includes durable aluminum diamond plate floor finish
  • Equipped with high-security steel or anodized aluminum doors
  • Offers R-10 wall insulation and R-19 ceiling insulation
  • Includes factory-installed lighting and electrical systems
  • Features a solar-reflective, waterproof roof membrane