Learn More
Med Vet International Guarded Culture Swab, J0272, 25-Pack
Catalog No. 5000582582
Encompass_Preferred
The most popularstyle of culture swab. A hinged guard or capcovers the tipped swab. This bullet-shaped tip allows easier insertion and protects the swab from contaminants. 28"L flexible rod compares in style to Kalayjian™ brand culture swabs.
Catalog No. 50-005-82582 Supplier Med Vet International Supplier No. JOR272
The most popularstyle of culture swab. A hinged guard or capcovers the tipped swab. This bullet-shaped tip allows easier insertion and protects the swab from contaminants. 28"L flexible rod compares in style to Kalayjian™ brand culture swabs.