Learn More
International Medical Industries (IMI) Guarded Luer Connector (Female Luer lock - Female Luer Lock) (Individual Blisters)
Catalog No. NC1758136
Encompass
Prep-Fill™ Guarded Luer Connectors (Female Luer lock - Female Luer Lock) provide convenient, high-quality solutions to facilitate transferring or mixing of fluids between drug delivery devices or IV sets. Prep-Fill™ Guarded Luer Connectors introduce feature enhancements to essential pharmaceutical compounding devices such as expanded wing size for secure manipulation, notched wing design that enhances first air over the connection port, and built-in drip tray design to capture stray fluid droplets during repeater pump procedures. • Sterile •Non-Pryogenic •Individual Blister •Box of 50
This item is not returnable. View return policy
Catalog No. NC1758136 Supplier International Medical Industries (IMI) Supplier No. 57400
Prep-Fill™ Guarded Luer Connectors (Female Luer lock - Female Luer Lock) provide convenient, high-quality solutions to facilitate transferring or mixing of fluids between drug delivery devices or IV sets. Prep-Fill™ Guarded Luer Connectors introduce feature enhancements to essential pharmaceutical compounding devices such as expanded wing size for secure manipulation, notched wing design that enhances first air over the connection port, and built-in drip tray design to capture stray fluid droplets during repeater pump procedures. • Sterile •Non-Pryogenic •Individual Blister •Box of 50