Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Guardian™ 2000 Hotplate and Stirrer

Catalog No. 01918390 Shop All OHAUS Products
Change view
Click to view available options
Temperature (Metric):
50°C to 380°C
70°C to 500°C
Size:
135 mm dia.
4 x 4 in.
7 x 7 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature (Metric) Size
01-918-390 50°C to 380°C 135 mm dia.
01-918-388 70°C to 500°C 4 x 4 in.
01-918-389 70°C to 500°C 7 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-918-390 Supplier OHAUS™ Supplier No. 30680257
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Guardian™ 2000 Hotplate and Stirrer is ideal for education and other lab environments that require affordable, everyday fast heating and reliable stirring up to 15 L. All three sizes offer simple knob control to easily adjust the heating and stirring functions.

  • Applications: Boiling Liquids, Basic Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
  • Display: Prominent hot top indicator light, green indicator lights when unit is heating and stirring
  • Operation: Simple knob control, stirring up to 1600 rpm, temperature up to 500°C (ceramic top) or 380°C (ceramic coated stainless steel top), power cord (included), stirring models include PTFE coated stir bar
  • Communication: N/A
  • Construction: Durable metal housing, PET front panel, control knobs, ON/OFF switch for heater disconnect, built in support rod holder, easy to clean resin, ceramic or ceramic coated stainless steel tops
  • Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring
TRUSTED_SUSTAINABILITY

Specifications

Top Plate Material Ceramic-coated Stainless Steel
Voltage 115 V
Controller Type Analog
Model e-G21HSRDS
Size 135 mm dia.
Temperature (Metric) 50°C to 380°C
Temperature (English) 122°F to 716°F
Frequency 60 Hz
Wattage 535 W
Weight (English) 5.5 lb.
Weight (Metric) 2.5 kg
Type Stirring Hotplate
Shape Round
Max. Sample Load 15 L
Plug Type US
No. of Positions 1
Max. Container Size 15 L
Stirring Range 200 to 1600 rpm
Warranty 2 Years
Dimensions (L x W x H) 264 x 163 x 101 mm (10.4 x 6.4 x 4 in.)
Display Analog
Speed 200 to 1600 rpm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.