Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Guardian™ 3000 Hotplate and Stirrer

Catalog No. 01918394 Shop All OHAUS Products
Change view
Click to view available options
Top Plate Material:
Ceramic
Ceramic-coated Stainless Steel
Temperature (Metric):
Ambient +5° to 380°C
Ambient +5° to 500°C
Size:
135 mm dia.
4 x 4 in.
7 x 7 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Top Plate Material Temperature (Metric) Size
01-918-394 Ceramic Ambient +5° to 500°C 4 x 4 in.
01-918-396 Ceramic-coated Stainless Steel Ambient +5° to 380°C 135 mm dia.
01-918-395 Ceramic Ambient +5° to 500°C 7 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-918-394 Supplier OHAUS™ Supplier No. 30680268
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Guardian™ 3000 Hotplate and Stirrer is designed for economical heating and stirring in all laboratory settings. Select from three plate sizes to suit your application. All models can be used with the optional temperature probe for precise temperature control of the sample.

  • Applications: Boiling Liquids, Water/Oil Bath Preparation, Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
  • Display: Backlit LCD Display for temperature and speed, green indicator lights when unit is heating and stirring, prominent hot top indicator light, probe in use icon, Single Point Calibration icon
  • Operation: Stirring from 80 to 1600 rpm, ambient temperature range from +5°C to 500°C (ceramic top) or 380°C (ceramic-coated steel top), power cord (included), stirring models include PTFE-coated stir bar
  • Communication: N/A
  • Construction: Durable metal housing, PET front panel, control knobs, on/off switch for heater disconnect, built-in support rod holder, easy-to-clean ceramic or ceramic-coated stainless steel tops
  • Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring, temperature probe connector for use with optional temperature probe
TRUSTED_SUSTAINABILITY

Specifications

Top Plate Material Ceramic
Voltage 115 V
Controller Type Digital
Model e-G31HS04C
Size 4 x 4 in.
Temperature (Metric) Ambient +5° to 500°C
Temperature (English) 41°F to 932°F
Frequency 50/60 Hz
Amperage 5 A
Wattage 550 W
Weight (English) 5.5 lb.
Weight (Metric) 2.5 kg
Type Stirring Hotplate
Shape Square
Max. Sample Load 15 L
Plug Type US
No. of Positions 1
Max. Container Size 15 L
Stirring Range 80 to 1600 rpm
Warranty 2 Years
Dimensions (L x W x H) 264 x 163 x 110 mm (10.4 x 6.4 x 4.3 in.)
Display Custom Backlit LCD
Speed 80 to 1600 rpm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.