Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guardian™ CP-13-11, Chemical Resistant Smooth Finish 13 mil Butyl Gloves
GSA_VASDP

Catalog No. 19321189
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-321-189 X-Small
19-321-190 Small
19-321-191 Medium
19-321-192 Large
19-321-193 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-321-189 Supplier Guardian™ Supplier No. 51501
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Butyl gloves offer protection against variety of aggressive/hazardous threats from chemicals, biologics, and radioactive materials. Ergonomic curved hand style is comfortable, with a smooth finish for enhanced grip capabilities.

  • For use with aldehydes, ketones, esters, and concentrated mineral acids
  • Resistant to oxidation, ozone corrosion, and abrasion
  • Shorter, thinner format than other Guardian™ Butyl Gloves
  • Rinse clean and hang dry for reuse
  • Made in the USA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Color Black
Latex Free Yes
Material Butyl
Size X-Small
Thickness (English) 13 mil
Thickness (Metric) 0.33 mm
Glove Cuff Style Beaded
Grip Design Smooth
Lining Unlined
Model 51501
Length (English) 11 in.
Length (Metric) 27.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.