Learn More
MSC Guardian Portable Eye Wash, 15 gal., Drench Hose, 1 ea
Catalog No. 5000793420
Encompass
This portable eye wash station has a 15-gal. capacity, designed to deliver a flow rate of 0.65 GPM at 80 PSI. It includes a drench hose for added functionality.
  • Pressurized for immediate use
  • Portable design for flexible placement
  • Tank capacity of 15 gal. (56 L)
  • Maximum flow rate of 0.7 GPM
  • Constructed from stainless steel
  • Includes ANSI-compliant sign
  • ANSI certified
Catalog No. 50-007-93420 Supplier MSC Supplier No. 69838431
This portable eye wash station has a 15-gal. capacity, designed to deliver a flow rate of 0.65 GPM at 80 PSI. It includes a drench hose for added functionality.
  • Pressurized for immediate use
  • Portable design for flexible placement
  • Tank capacity of 15 gal. (56 L)
  • Maximum flow rate of 0.7 GPM
  • Constructed from stainless steel
  • Includes ANSI-compliant sign
  • ANSI certified