Learn More
Glass Expansion Probe Connecting Line 1.0mm ID (Green)
SDP
Catalog No. NC2470739
Encompass_Preferred
Probe Connecting Line 1.0mm ID (Green). Compatible with Glass Expansion Guardian Probes for Aim Lab & Agilent SPS3/SPS4 Autosamplers (P/N 70-803-1957) and for Teledyne Cetac autosamplers (P/N 70-803-1803). Creates a zero dead-volume connection between Guardian Probe and nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900
Catalog No. NC2470739 Supplier Glass Expansion Supplier No. 708031721
Probe Connecting Line 1.0mm ID (Green). Compatible with Glass Expansion Guardian Probes for Aim Lab & Agilent SPS3/SPS4 Autosamplers (P/N 70-803-1957) and for Teledyne Cetac autosamplers (P/N 70-803-1803). Creates a zero dead-volume connection between Guardian Probe and nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900