Learn More
Glass Expansion Guardian Probe for Cetac ASX-112FR
SDP
Catalog No. 502703022
Encompass_Preferred
Guardian Probe for ASX-112FR, probe only. Compatible with Teledyne Cetac ASX-112FR autosampler. Requires a UniFit sample capillary available in IDs of 0.18mm (P/N 70-803-2085), or 0.25mm (P/N 70-803-2030). The unique, robust tip design eliminates crushed and damaged tips due to misalignment. Drip-resistance prevents cross contamination of samples during probe movement, especially with oils. Built-in particle filtering holds back particulates from blocking the line. Completely inert design, all PEEK and PTFE construction, the Guardian autosampler probe provides exceptional resistance to strong acids and solvents.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900
Catalog No. 50-270-3022 Supplier Glass Expansion Supplier No. 708032029
Guardian Probe for ASX-112FR, probe only. Compatible with Teledyne Cetac ASX-112FR autosampler. Requires a UniFit sample capillary available in IDs of 0.18mm (P/N 70-803-2085), or 0.25mm (P/N 70-803-2030). The unique, robust tip design eliminates crushed and damaged tips due to misalignment. Drip-resistance prevents cross contamination of samples during probe movement, especially with oils. Built-in particle filtering holds back particulates from blocking the line. Completely inert design, all PEEK and PTFE construction, the Guardian autosampler probe provides exceptional resistance to strong acids and solvents.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900