Learn More
Glass Expansion Guardian Probe for Cetac ASX-200/500/800 Series. Guardian probe only.
SDP
Catalog No. 502703036
Encompass_Preferred
Guardian Probe Cetac ASX-200/500/800 Series, Guardian Probe Only. The unique, robust tip design eliminates crushed and damaged tips due to misalignment. Drip-resistance prevents cross contamination of samples during probe movement, especially with oils. Built-in particle filtering holds back particulates from blocking the line. Completely inert design, all PEEK and PTFE construction, the Guardian autosampler probe provides exceptional resistance to strong acids and solvents. The Guardian probe has an ID of 1.0mm, with interchangeable UniFit sample capillaries available in IDs of 0.3mm, Black (P/N 70-803-1853), 0.5mm, Blue (P/N 70-803-1852), 0.75mm, Red (P/N 70-803-1714) and 1.0mm, Green (P/N 70-803-1721).Guardian Probe for Aim Lab & Agilent SPS3/SPS4 Autosamplers, see P/N 70-803-1957.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900
Catalog No. 50-270-3036 Supplier Glass Expansion Supplier No. 708031787
Add to Cart
Add to Cart
Guardian Probe Cetac ASX-200/500/800 Series, Guardian Probe Only. The unique, robust tip design eliminates crushed and damaged tips due to misalignment. Drip-resistance prevents cross contamination of samples during probe movement, especially with oils. Built-in particle filtering holds back particulates from blocking the line. Completely inert design, all PEEK and PTFE construction, the Guardian autosampler probe provides exceptional resistance to strong acids and solvents. The Guardian probe has an ID of 1.0mm, with interchangeable UniFit sample capillaries available in IDs of 0.3mm, Black (P/N 70-803-1853), 0.5mm, Blue (P/N 70-803-1852), 0.75mm, Red (P/N 70-803-1714) and 1.0mm, Green (P/N 70-803-1721).Guardian Probe for Aim Lab & Agilent SPS3/SPS4 Autosamplers, see P/N 70-803-1957.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-900