Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guardian Protective Wear Hoods

Catalog No. p-3638094
Change view
Click to view available options
Color:
Lime Yellow
Olive Drab
Yellow
Includes:
Drawstring
Waterproof seal, snap for attaching to collar
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
19-047731 Yellow Waterproof seal, snap for attaching to collar
19-812-247 - Drawstring
19-812-222 Lime Yellow Waterproof seal, snap for attaching to collar
19-812-223 Olive Drab Waterproof seal, snap for attaching to collar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-047731 Supplier Guardian Protective Wear Supplier No. 985Y
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Size Universal
Color Yellow
Seam Type Sewn and taped
Quantity 1 Ea.
Includes Waterproof seal, snap for attaching to collar
For Use With (Application) Wet environment
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.