Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guardian Protective Wear Tri-Weave Overalls

Catalog No. 19812146
Change view
Click to view available options
Size:
Medium
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-146 2X-Large
19-812-144 Medium
19-812-145 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-812-146 Supplier Guardian Protective Wear Supplier No. 706Y2XLI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Closure Type Adjustable Suspenders
Color Yellow
Material Nylon, Vinyl
Seam Type Stitched, Taped
Size 2X-Large
Construction Tri-weave
For Use With (Application) Protection from chemicals
Style Bib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.