Learn More
Mopec Guardian Rack System
SDP
Catalog No. 22445163 Shop All Mopec Products
Change view
Click to view available options
Tiers:
3
4
5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tiers
22-445-163 3
22-445-164 4
22-445-165 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-445-163 Supplier Mopec Supplier No. IR26301
Add to Cart
Add to Cart
Collapsible morgue storage rack system
  • Easily to assemble and disassemble – only 8 bolts
  • Heavy duty design – Holds up to 1,500 lbs in body weight and lasts for many years and many assemble/disassemble cycles
  • Suitable for refrigerated trailers or other cooled structures
  • 3, 4 and 5 tier designs
  • Large, heavy-duty casters are easy to roll and lockable
  • Designed for loading using a forklift
  • All stainless steel construction
Specifications
Material Stainless Steel
Tiers 3
Width (English) 26 in.