Learn More
MSC Guardian Rain Bib Pant, Yellow, Medium, 1 ea
Catalog No. 5000792240
Encompass
The Guardian Rain Bib Pant provides reliable splash protection from most chemicals. Constructed with Vinyl/Nylon/Vinyl material, it features seams that are sewn and tape-sealed, and it is flame retardant to meet NFPA 701 standards.
  • High abrasion and tear resistance
  • Flexible at temperature extremes
  • Non-conductive nylon straps with no metal parts
  • Easy care and maintenance
  • Guaranteed against defects in material or workmanship
Catalog No. 50-007-92240 Supplier MSC Supplier No. 91979047
The Guardian Rain Bib Pant provides reliable splash protection from most chemicals. Constructed with Vinyl/Nylon/Vinyl material, it features seams that are sewn and tape-sealed, and it is flame retardant to meet NFPA 701 standards.
  • High abrasion and tear resistance
  • Flexible at temperature extremes
  • Non-conductive nylon straps with no metal parts
  • Easy care and maintenance
  • Guaranteed against defects in material or workmanship