Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guardian Protective Wear Rainwear Coveralls

Catalog No. 19812153
Change view
Click to view available options
Color:
Green
Yellow
Size:
Small
X-Large
2X-Large
3X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-812-153 Green 2X-Large
19-812-152 Green X-Large
19-812-160 Yellow 3X-Large
19-812-159 Yellow Small
19-812-156 Yellow X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-153 Supplier Guardian Protective Wear Supplier No. 756GN2XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Size 2X-Large
Material Vinyl
Color Green
Seam Type Taped
Certifications/Compliance NFPA 701
Size (English) Waist 33 in.
Size (Metric) Waist 83.82 cm
Waist Style Adjustable drawstring
For Use With (Application) Splash protection from most chemicals
Item Description Adjustable drawstring waist, plain front
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.