Learn More
Aviva Systems Biology Corp GUCA1B Antibody - N-terminal region (ARP59766_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59766_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GUCA1B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GUCA1B. Alternate names: DKFZp686E1183, GCAP2, GUCA2, RP48ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-817